You Searched For: Microplate Shakers
19  results were found
SearchResultCount:"19"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (444-0270)
Supplier: VWR Collection
Description: Microplate shaker with microprocessor control which holds two or four microplates, deep well plates or two microtube racks. Variable speed across a wi...
UOM: 1 * 1 items
Waste Reducing
Catalog Number: (444-7016)
Supplier: VWR Collection
Description: Ideal for molecular biology applications, mechanical and chemical cell lysis, mixing tissue samples, mixing cytogenetic suspensions, and vortexing cel...
UOM: 1 * 1 items
Waste Reducing
Supplier: Heidolph Instruments GmbH & Co.KG
Description: For vigorous mixing of microtitre plates. Vibration orbit of 1,5 mm guarantees no spills even with full vessels, 3 mm orbit gives best mixing results ...
Catalog Number: (444-0024)
Supplier: IKA
Description: Microplate shaker with electronic control keeps the set speed constant via rated/actual speed comparison. Supplied with attachment but without microti...
UOM: 1 * 1 items
Catalog Number: (SCIISI-4102)
Supplier: SCIENTIFIC INDUSTRIES (ELMIS)
Description: Fully programmable microplate shaker for four plates (8 with optional tray).
UOM: 1 * 1 items
New Product
Catalog Number: (444-0041)
Supplier: GRANT INSTRUMENTS LTD.
Description: Compact microplate shaker which provides reliable and regulated shaking for 96- and 384-well microplates with easy set up and digital timer. Variable ...
UOM: 1 * 1 items
Catalog Number: (444-9441)
Supplier: Edmund Bühler GmbH
Description: The TiMix 2 microplate shaker is specially for small samples in microplates, deepwell-plates or other plates in microplate format.
UOM: 1 * 1 items
Supplier: Edmund Bühler GmbH
Description: High capacity microplate shaker with high speed orbital motion. TiMix 5 is an analogue version, and TiMix 5 control has digital display and programmab...
Catalog Number: (EPPE5353000.510)
Supplier: EPPENDORF
Description: Innovative, flexible and compact, this mixer is specifically designed for mixing small volumes in numerous plate and tube formats. Rapid and precisely...
UOM: 1 * 1 items
Catalog Number: (BOEI130000-2)
Supplier: BOEKEL SCIENTIFIC
Description: The Jitterbug microplate shaker is ideal for immunoassays like ELISA, PCR and other research procedures requiring heating and shaking of microplates.
UOM: 1 * 1 items
Supplier: BOEKEL SCIENTIFIC
Description: Designed to suitable for two or four standard microplates or strip trays.
Catalog Number: (OHAU30391907)
Supplier: OHAUS
Description: This light duty microplate shaker is ideal for immunoassays and general microplate shaking, and is designed to be used in a variety of environmental c...
UOM: 1 * 1 items
Supplier: Cole Parmer
Description: Shaker and incubator combination makes these devices ideal for cell culture procedures.
Catalog Number: (STUA79520-22)
Supplier: Cole Parmer
Description: Combine with a recirculating cooler for low-temperature applications.
UOM: 1 * 1 items
Catalog Number: (BOEI130000)
Supplier: BOEKEL SCIENTIFIC
Description: Quiet, rugged shaker is ideal for a variety of immunoassay and research applications.
UOM: 1 * 1 items
Supplier: H + P LABORTECHNIK
Description: Ultra flat microplate shakers with VARIOMAG drive which are ideally suited for easy integration into robotic systems. Monoshake is a directly controll...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 19
no targeter for Bottom