You Searched For: Water Samplers
10  results were found
SearchResultCount:"10"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Hach
Description: These lightweight samplers with low power requirements (12 VDC) have been designed for easy mobility.
Supplier: BÃœRKLE
Description: Samplers available in three sizes.
Catalog Number: (705-0989)
Supplier: Hach
Description: Portable water sampler using pressure-vacuum technology. Combines small dimensions by offering the 24×1 l bottle option for glass as well as plastic b...
UOM: 1 * 1 items
Supplier: MAXX
Description: The SP 5 are modular fixed site units available in different configurations.
Supplier: MAXX
Description: Water samplers with thermostatic control and protective top, which can be opened for connection and maintenance works. Self-contained, automatic cooli...
Supplier: MAXX
Description: A portable, battery operated, and compact automated water sampler with a diaphragm pump vacuum collection system. The standard version has an integrat...
Supplier: Sampling Systems
Description: The Deep water sampler can take samples of liquids from large depths.
New Product
Catalog Number: (701-0166)
Supplier: MAXX
Description: Wall-mounted sampling device with housing made from Styrosun/PC (GF10). No thermostating.
UOM: 1 * 1 items
Catalog Number: (MAXA2070001)
Supplier: MAXX
Description: This portable water sampler is a unique combination of design and technology as well as an unrivaled measuring device for volume determination.
UOM: 1 * 1 items
Supplier: BEHR
Description: The sampling scoop is composed of a clamping fixture and end bar. The scoop with receptacles made of borosilicate glass comply with the high requireme...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 10 of 10
no targeter for Bottom