You Searched For: Solid Samplers
73  results were found
SearchResultCount:"73"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (300-0162)
Supplier: VWR Collection
Description: Strong plastic briefcase with various sampling tools for food products, animal feed and water.
UOM: 1 * 1 items
Supplier: BÃœRKLE
Description: For sampling bulk goods, cross-sectional samples can be taken from different sample depths. Suitable for use with a range of sample substrates from fi...
Supplier: BÃœRKLE
Description: When bulk goods are transported, they may separate out and surface samples may not be representative. The sampler is opened manually once it has reach...
Supplier: BÃœRKLE
Description: Conical hand drill with sharp cutting edges and a solid handle made from stainless steel V2A (1.4301)/AISI 304.
Catalog Number: (BURK5310-0150)
Supplier: BÃœRKLE
Description: This sampler with drilling blades makes it easy to take samples of media, even those with high density, and every type of highly compacted powders and...
UOM: 1 * 1 items
Supplier: BÃœRKLE
Description: Guarantees precise zone sampling through all layers of the sample. The spiral arrangement of the sampling slits means that these are opened one after ...
Supplier: BÃœRKLE
Description: Novartos Uno sampler with two or three sample chambers, for one sample zone.
Sale
Supplier: BÃœRKLE
Description: Novartos is the pharmaceutical zone sampler for sampling in accordance with the FDA guidelines for unit dose sampling. With the Novartos pharmaceutica...
Supplier: BÃœRKLE
Description: Stainless steel AISI 316 L (1,4404). The MicroSampler has been specially designed for the sampling of small quantities of powder.
Supplier: BÃœRKLE
Description: The sample drill for stalk material is a sampler especially for hay, silage, straw, herbs, bushes or other stalky plant dry material where other drill...
New Product
Supplier: 3M Food Safety
Description: 3Mâ„¢ Swab-sampler with Letheen broth is a convenient and quick collection device for environmental sampling and testing over small surface areas.
Supplier: BÃœRKLE
Description: Quality assurance starts with the inspection of incoming goods. An all-layer sample of bulk goods is no longer a problem with PowderProof®, the sample...
Supplier: BÃœRKLE
Description: Stainless steel sampler for powders. The conical inlet allows for collection of different sample sizes, up to approximately 50 ml. Constructed without...
Supplier: BÃœRKLE
Description: Sampler for free flowing bulk goods up to approx. 10 mm diameter. This makes it possible, for example, to remove hazel nuts, blends of tea, grain or s...
Supplier: BÃœRKLE
Description: Made of stainless steel or PP, transparent, sample bottles made of PE. The QuickPicker is appropriate when bulk goods are withdrawn directly from sack...
Supplier: BÃœRKLE
Description: To make a sampling procedure reproducible, it should always be carried out under the same conditions. For this purpose, the depth stop for samplers is...
New Product
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 73
no targeter for Bottom