You Searched For: Roller Culture Apparatus
6  results were found
SearchResultCount:"6"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (734-1167)
Supplier: DWK Life Sciences
Description: This conventional type roller culture equipment is designed to roll vessels 108 to 121 mm in Ø and up to 290 mm long.
UOM: 1 * 1 items
Supplier: DWK Life Sciences
Description: The Wheaton® R2P™ 2.0 roller culture apparatus is designed to allow for the development and growth of cultured cells from research to production quant...
Supplier: DWK Life Sciences
Description: This standard cell production roller culture apparatus allows flexibility for scale-up and production of monolayer cell cultures in standard roller bo...
Catalog Number: (734-1164)
Supplier: DWK Life Sciences
Description: The mini bottle bench top roller culture system is designed for small scale mixing and agitation using bottles that are too small to be accommodated b...
UOM: 1 * 1 items
Supplier: BELLCO GLASS INC.
Description: Roller culture apparatus, Modular with all-position drive, fixed frame, stand-by drive and alarm, Sci/ERAâ„¢, 45 bottles (9 decks)
Supplier: BELLCO GLASS INC.
Description: Roller culture apparatus, Modular, 25 bottles (4 decks)
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 6 of 6
no targeter for Bottom