You Searched For: Benchtop Refractometers
20  results were found
SearchResultCount:"20"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Simple to use, precise bench top refractometers for use in laboratory or production environments.
Supplier: BELLINGHAM & STANLEY
Description: Robust fully automatic instruments for the food, sugar and beverage industries where temperature control is not required, such as measuring sugar (°Br...
New Product
Supplier: BELLINGHAM & STANLEY
Description: These high precision refractometers are designed for applications in food and beverage, including fruit juice, emulsions or chemicals.
New Product
Catalog Number: (BELL19-20)
Supplier: BELLINGHAM & STANLEY
Description: The RFM742-M refractometers are robust fully automatic instruments suited to the food, sugar and beverage industries where temperature control is not ...
UOM: 1 * 1 items
New Product
Supplier: BELLINGHAM & STANLEY
Description: RFM900-T are suitable for chemical, petrochemical, pharmaceutical, flavours and fragrance industries, as well as for academic research.
Supplier: BELLINGHAM & STANLEY
Description: These refractometers bring with a line of high precision Peltier temperature controlled benchtop refractometers for demanding applications within the ...
New Product
Catalog Number: (635-0480)
Supplier: BELLINGHAM & STANLEY
Description: The Abbe refractometer is designed for applications in educational establishments and laboratories, where wide refractive index ranges are required, o...
UOM: 1 * 1 items
Supplier: ATAGO
Description: The RX-5000α are automatic digital refractometers, which can set measurement temperature internally and have the ability to measure the refractive ind...
Catalog Number: (635-0148)
Supplier: ATAGO
Description: The RX-7000α is an automatic digital refractometer, which can set measurement temperature internally and measure the refractive index, °Brix or concen...
UOM: 1 * 1 items
Supplier: ATAGO
Description: The NAR Abbe refractometers can meet many of the requirements for routine measurements.
Catalog Number: (ATAG3921)
Supplier: ATAGO
Description: The RX-007α is designed for water soluble samples with very low concentrations (5000 % or less) such as tea and low sugar beverages at a very high acc...
UOM: 1 * 1 items
Supplier: ATAGO
Description: These multi-wavelength Abbe refractometers can measure refractive index or Abbe number ([nu]d or [nu]e) at different wavelengths.
Supplier: ATAGO
Description: Accurate digital refractometers featuring intuitive colour touch screen technology makes the RX-i range easy to use and navigate around the operating ...
Catalog Number: (ATAG3150)
Supplier: ATAGO
Description: This refractometer is designed for the determination of sugar levels in beverage, food and pharmaceuticals.
UOM: 1 * 1 items
Supplier: ATAGO
Description: These Abbe refractometers are designed to meet the requirements in routine applications.
Catalog Number: (635-0397)
Supplier: ATAGO
Description: The RX-9000α is an automatic digital refractometer, which can set measurement temperature internally and measure the refractive index, Brix degrees or...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 20
no targeter for Bottom