Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (13)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "peristaltic-pumps"
https://ru.vwr.com/stibo/smallweb/std.lang.all/96/38/18989638.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"38"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   The VWR FASTLoad peristaltic pump series provides highly accurate and reproducible flow rates from 0,8 up to 1312 ml/min.
Supplier:  VWR Collection
Description:   PP 3300D and PP 3400D peristaltic pumps are specifically designed for critical metering and dispensing applications: The user can pump, dispense and f...
Supplier:  VWR Collection
Description:   The PP 4000 series peristaltic pumps provide multichannel pumping with the accuracy of flow control and broad flow range to efficiently service most p...
Supplier:  VWR Collection
Description:   The VWR® PP 10 and PP 22 pumps offer enhanced and versatile performance in an ultra-compact, low maintenance design. These highly innovative peristalt...
Supplier:  VWR Collection
Description:   VWR PP 3300 and PP 3400 peristaltic pumps are ideal general purpose tubing pumps for high repeatability, precision metering, and worry-free performanc...
Catalog Number: (18-1110-91)
Supplier:  GE Healthcare - Life Sciences
Description:   Single channel laboratory pump for use in liquid chromatography and other applications where accurately
controlled liquid flows are required.
UOM:  1 * 1 items
Catalog Number: (516-0952)
Supplier:  Pall Laboratory
Description:   The Sentino® pump system is ideal for microbiological MF technique using Pall MicroFunnel™ filtration units. Designed to use peristaltic action to dra...
UOM:  1 * 1 items
Supplier:  ISMATEC PART OF COLE PARMER
Description:   Peristaltic pump, ECOLINE VC-380, Channels: 1, Flow rate: 1,6 - 5000 ml/min, W×D×H: 169×256×138 mm, Weight: 5,3 kg
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 38
  1  2  3  4  5  Next