Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "petri-dish-fillers"
https://ru.vwr.com/stibo/smallweb/std.lang.all/96/42/18989642.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"6"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (PBIB13120)
Supplier:  VWR Collection
Description:   This compact instrument from stainless steel dispenses medium automatically to Petri dishes.
UOM:  1 * 1 items
Supplier:  BIOTOOL AG
Description:   PetriSwiss PS 20 fills 20 dishes directly in the PetriRack. There is no need for manual stacking or any other manual operations. It is ideal for produ...
Supplier:  BIOTOOL AG
Description:   PetriSwiss PS 200 and 400 automatically fill and stack Petri dishes in removable safety racks. The unit takes the lowest dish from the stack and place...
Supplier:  BIOTOOL AG
Description:   TubeFiller was designed to reduce repetitive manual filling of any format laboratory racked tubes. Based on its dedicated function to fill “any tubes ...
Supplier:  BIOTOOL AG
Description:   The Petri Dish filling solution designed for high throughput. All entries are made directly on the screen! No manual is needed. Operator promting in G...
Supplier:  INTEGRA
Description:   The Mediajet Vario is the perfect filler for a wide range of Petri dish sizes; it can be adapted to fill Petri dishes of 60 or 90 mm diameters in...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 6  of 6