You Searched For: Vacuum Ovens
5  results were found
SearchResultCount:"5"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: MEMMERT
Description: Reliable, powerful, energy efficient vacuum ovens with separately adjustable thermo shelves which are directly heated ensuring precise temperature con...
Supplier: Binder
Description: The VD series of vacuum drying chambers provide fast gentle drying of non flammable solvents, delivering impressive performance through their precise ...
Supplier: Thermo Scientific
Description: Vacuum drying ovens with stainless steel chambers, vacuum fittings, tubing and ball valve. Units are designed for gentle drying of heat-sensitive samp...
Supplier: BARNSTEAD (INC THERMOLYNE)
Description: Achieve maximum temperatures of 220 °C with these Thermo Scientific™ vacuum ovens, while benefiting from two control configurations and display option...
Catalog Number: (METN51371200)
Supplier: Mettler - Toledo
Description: The DO308 drying oven is the ideal accessory for the METTLER TOLEDO volumetric or coulometric Karl Fischer titrators. It allows easy and accurate wate...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 5 of 5
no targeter for Bottom