You Searched For: Microwave Ovens
5  results were found
SearchResultCount:"5"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: SMEG S.P.A.
Description: Microwave oven universale stainless steel have freestanding microwave oven with grill.
Supplier: CEM
Description: Microwave oven, MARSXpress Vessel Starter Set, PFA, 40 place, 55 ml
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Description: Stainless steel exterior and interior.
Catalog Number: (CEMA920085)
Supplier: CEM
Description: SAM-255 with IntelliTempâ„¢ infrared temperature control provides rapid drying of all types of samples. Workstation can analyse large sized samples for ...
UOM: 1 * 1 items
Catalog Number: (CEMAS-924560)
Supplier: CEM
Description: This rapid microwave ashing system consists of a microwave heating system containing a 5 L chamber which can be heated to 1000 °C or a 1,8 L chamber w...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 5 of 5
no targeter for Bottom