Hybridisation ovens, Hybaid Shake ‘n' Stack
Supplier: Thermo Fisher Scientific
Hybaid
732-1835EA 4781 EUR
732-1835 732-1836
Hybridisation ovens, Hybaid Shake ‘n' Stack
Ovens Hybridisation Ovens
The Hybaid Shake ‘n' Stack has been specifically designed for safe stacking up to three units high. Each oven has the ability to operate with either a carousel or shaking platform. One compact triple oven tower can be set up to undertake three distinct functions at different temperatures. Each oven can be furnished with a range of accessories, including a variety of carousels.
  • Multiple carousel fittings for flexible choice of consumables – able to hold up to ten 35 mm diameter hybridisation bottles or thirty 50 ml plastic tubes
  • Interchangeable carousel/shaking platform for hybridisation and washing procedures
  • Stackable format allowing better utilisation of laboratory space
  • Variable speed settings for protocol optimisation
Variable axis carousel
Variable speed carousel
Delivery information: Hybaid Shake 'n' Stack is supplied with or without shaker platform. All models include ten 35 mm bottle capacity plastic carousel, adjustable feet, drip tray and manual.