VWR® DRY-Line®, Ovens with Natural Convection
Supplier: VWR
VWR®
466-3513EA 0 EUR
466-3513 466-3510
VWR® DRY-Line®, Ovens with Natural Convection
Ovens Standard Ovens
Drying ovens with natural convection used for thermal processes, heated storage and for convection drying applications with temperatures up to 220 °C. Exhaust duct at the back of the unit with manually adjustable slide.
  • Microprocessor controller
  • Integrated timer can be set for continuous mode or timer operation: 0 to 999 min or 0 to 99,9 h
  • Over-temperature protection with visual alarm (Class 3.1 safety device)
  • Temperature setting in increments of 1 °C
Delivery information: Supplied with two chrome plated shelves.