You Searched For: Ovens
62  results were found
SearchResultCount:"62"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: VENTI-Line® drying ovens are designed for all standard drying and tempering tasks as well as sterilising glassware. They have a temperature range betw...
Supplier: VWR Collection
Description: DRY-Line® drying ovens are designed for all standard drying and tempering tasks as well as sterilising glassware. They have a temperature range betwee...
Sale
Catalog Number: (732-2956)
Supplier: VWR Peqlab
Description: A flexible and cost effective platform for any laboratory, the PerfectBlot can accommodate multiple 150 mm or 300 mm bottles at temperatures up to 80 ...
UOM: 1 * 1 items
Catalog Number: (455-1100)
Supplier: NABERTHERM GMBH
Description: The L 3/11 - L 40/11 muffle furnaces are the ideal choice for daily laboratory use. These high quality models have an advanced and attractive design, ...
UOM: 1 * 1 items
Supplier: NABERTHERM GMBH
Description: The durability of the SiC rods in periodic use, in combination with their high heating speed, make these high-temperature furnaces good all-rounders i...
Supplier: NABERTHERM GMBH
Description: These compact bench top tube furnaces with integrated controllers can be widely used for a variety of processes. Equipped with a standard C 530 cerami...
Supplier: MEMMERT
Description: UNpa paraffin ovens provide precise and homogenous temperature control of paraffin embedding medium without any loss in quality. UNpa paraffin ovens a...
Supplier: MEMMERT
Description: The range includes natural convection models with a temperature range up to +300 Â°C. There is a choice of two different displays, SingleDISPLAY o...
Supplier: NABERTHERM GMBH
Description: These furnaces feature heating elements on supporting tubes that radiate freely into the chamber, ensuring particularly short heating times. With the ...
New Product
Supplier: NABERTHERM GMBH
Description: These furnaces feature heating elements on supporting tubes that radiate freely into the chamber, ensuring particularly short heating times. With the ...
New Product
Supplier: MEMMERT
Description: The range includes forced convection models with a temperature range up to 300 Â°C. There is a choice of two different displays, SingleDISPLAY or ...
Supplier: MEMMERT
Description: The UF TS pass through ovens help to minimise contamination when transporting samples or components. They have an easy-to-clean interior, fast recover...
Supplier: NABERTHERM GMBH
Description: These compact tube furnaces are used when laboratory experiments must be performed horizontally, vertically, or at specific angles. The ability to con...
Supplier: NABERTHERM GMBH
Description: With their convincing price/performance ratio and the fast heat-up rates, these compact muffle furnaces are perfect for many applications in the labor...
Catalog Number: (732-4301)
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: The HybriLinker oven combines an HB-1000 hybridisation oven with a CX-2000 crosslinker with operation independent of one another. The crosslinker port...
UOM: 1 * 1 items
Catalog Number: (732-4315)
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: The Minidizer oven is designed as a personal desk-top unit that is both compact and portable. It is ideal for use in start-up laboratories or those wi...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 62
no targeter for Bottom