Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (4)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Digital Cameras
https://ru.vwr.com/stibo/smallweb/std.lang.all/51/17/18945117.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"25"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (630-2671)
Supplier:  VWR Collection
Description:   CMOS camera with high speed output, built-in buffer 32 Mb memory and image transmission.
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   The VisiCam plus cameras offer high resolution high speed live imaging and fit any microscope in educational, industrial or clinical use.
Catalog Number: (630-2629)
Supplier:  VWR Collection
Description:   Wi-Fi CS-mount camera, VWR® VisiCam®, Resolution: 5,0 MP (PC) ; 2,0 MP (WiFi), CMOS, Exposure time: Automatic, Sensor: Micron C-MOS, 80 g
UOM:  1 * 1 items
Catalog Number: (630-2957)
Supplier:  VWR Collection
Description:   USB C-mount and eyepiece microscope camera.
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   These cameras provide live images and video capture to an SD card or to connect to a PC.
Catalog Number: (630-2669)
Supplier:  VWR Collection
Description:   Scientific CMOS camera to deliver high frame rate and stable image transmission.
UOM:  1 * 1 items
Catalog Number: (630-2668)
Supplier:  VWR Collection
Description:   Thanks to the built-in 16 MB buffer memory, it delivers high frame rate and stable image transmission.
UOM:  1 * 1 items
Supplier:  MOTIC
Description:   Digital camera, Moticam®, Moticam 1, Resolution: 800×600 P, Sensor: CMOS
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 25
  1  2  3  4  Next