You Searched For: Cold Light Sources
10  results were found
SearchResultCount:"10"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: The passively-cooled VisiLight® LED3 cold light source, with fanless cooling system, can be used for a variety of applications, such as quality assura...
Supplier: VWR Collection
Description: This cold light source with compact and stable aluminium housing and temperature fan control provides an optimal and homogenous illumination of the li...
Catalog Number: (631-1085)
Supplier: VWR Collection
Description: This high power 14 W LED source provides a pure-white light (6300 K).
UOM: 1 * 1 items
Supplier: VWR Collection
Description: This fanless cold light source with passive cooling system and compact and stable aluminium housing provides an optimal and homogenous illumination of...
Supplier: SCHOTT AG Lighting and imaging
Description: These LED cold light sources, for eco-friendly PURAVIS® light guides, are driven by a multiple LED module and provide a powerful illumination system f...
Catalog Number: (SCOC400350)
Supplier: SCHOTT AG Lighting and imaging
Description: Professional incident and darkfield illumination.
UOM: 1 * 1 items
Supplier: SCHOTT AG Lighting and imaging
Description: Professional illumination for stereo microscopy. The VisiLED product line offers modular, homogeneous, and shadow-free LED illumination for stereo mic...
Catalog Number: (630-2597)
Supplier: SCHOTT AG Lighting and imaging
Description: This professional halogen cold light source, for eco-friendly PURAVIS® glass fibre optic light guides, has been developed with the full 150 W halogen ...
UOM: 1 * 1 items
Catalog Number: (630-1528)
Supplier: SCHOTT AG Lighting and imaging
Description: This compact LED cold light source for eco-friendly PURAVIS® light guides is driven by a single LED and provides a brightness equivalent to a 30 W hal...
UOM: 1 * 1 items
Catalog Number: (EUMXLE5210)
Supplier: EUROMEX
Description: This popular multifunctional cold light source contains a 100 W halogen light source.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 10 of 10
no targeter for Bottom