You Searched For: Microscopes
133  results were found
SearchResultCount:"133"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: VisiScope 800 laboratory microscopes are designed for research projects.
Catalog Number: (630-1943)
Supplier: VWR Collection
Description: The VisiScope 350 microscopes with V-shaped Greenough optical system can be used for observations in advanced teaching, laboratories and industrial ap...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Mid-level microscopes for use in universities, laboratories and in industry.
Catalog Number: (630-1936)
Supplier: VWR Collection
Description: Upright microscope for brightfield and polarising light observations with strain-free IOS N-plan POL objectives. Perfect for accurate polarisation ana...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: VisiScope ALC microscopes have been designed to fulfil all the requirements of an advanced biology educational laboratory.
Supplier: VWR Collection
Description: Reliable, advanced microscopes for educational purposes. Particularly recommended for secondary schools.
Supplier: VWR Collection
Description: These monocular and binocular microscopes, with integrated camera, are ideal for educational purposes.
Supplier: VWR Collection
Description: Educational microscopes for transmitted light observations in routine applications.
Supplier: VWR Collection
Description: These polarising microscopes with rechargeable battery for very long outdoor operation (up to 15 hours) are ideal for educational purposes.
Supplier: VWR Collection
Description: Choose from a range of binocular and trinocular stereo zoom microscope heads for either basic or advanced requirements.
Supplier: VWR Collection
Description: Routine inverted fluorescence microscopes for transmitted brightfield, phase contrast (optional for IT407 FL4 and IT407 FLD4 FLD) and fluorescence obs...
New Product
Catalog Number: (630-2942)
Supplier: VWR Collection
Description: Binocular stereo microscope equipped with turnable objective (2× to 4×), fixed arm with handle, and bright, uniform LED illumination (transmitted and ...
UOM: 1 * 1 items
Catalog Number: (630-3290)
Supplier: VWR Collection
Description: Educational and laboratory microscope with integrated camera and removable tablet.
UOM: 1 * 1 items
Catalog Number: (630-1577)
Supplier: VWR Collection
Description: Reliable microscopes for educational purposes. Particularly recommended for secondary and higher schools. SZB250 and SZT250 have a 0,7× to 4,5× zoom r...
UOM: 1 * 1 items
Catalog Number: (630-2982)
Supplier: VWR Collection
Description: This compact Greenough stereo microscope with 6.72:1 zoom and 110 mm working distance is ideal for routine inspections in professional environments an...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: These upright microscopes are ideal for use in universities and routine laboratories.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 133
no targeter for Bottom