Microplate dispenser, Multidrop® DW
Supplier: Thermo Scientific
735-0441EA 8855 EUR
LABL5840170
Microplate dispenser, Multidrop® DW
The Multidrop® DW is a high-speed, automated dispenser designed for Deep Well plates, 1,1ml test tubes and 96-well plates. The instrument dispenses volumes of 20 to 995 µl in 5 µl increments.The Multidrop® DW can dispense up to eight different reagents with high accuracy, precision and speed. All reagent lines can be back-flushed to the reagent bottle, minimising the loss of expensive reagents. To ensure sterility and avoid cross-contamination, the dispensing cassette is detachable and autoclavable.
  • Superior accuracy and precision
  • Designed for repetitive dispensing of larger volumes
  • High Precision peristaltic pump gives high-speed continuous dispensing
  • Time controlled applications can be controlled by PC via RS232 serial interface.
  • Autoclavable dispensing cassette
The Multidrop® DW can be easily integrated into robotic systems.
  • Superior accuracy and precision
  • Designed for repetitive dispensing of larger volumes
  • High Precision peristaltic pump gives high-speed continuous dispensing
  • Time controlled applications can be controlled by PC via RS232 serial interface.
  • Autoclavable dispensing cassette
The Multidrop® DW can be easily integrated into robotic systems.
Ordering information: For details of accessories for the Multidrop® DW please see entry for Multidrop® 384 in this catalogue or on the VWR website www.vwr.com.