Melting point apparatus, analogue, SMP11
Supplier: Cole Parmer
stuart®
621-0036EA 1020 EUR
621-0036
Melting point apparatus, analogue, SMP11
Melting Point Apparatus
Samples in a capillary tube with sealed end are positioned in the heating block. The heating rate is adjusted manually. The instrument will rapidly heat up at a rate of 20 °C per minute to the set melt temperature and up to a maximum temperature of 250 °C. Accurate readings to within 1 °C of the melt temperature can be achieved by using a slower heating rate of between 1 and 10 °C per minute. Samples are illuminated by a bright white LED and viewed via a magnifying lens, which can be easily cleaned.
  • Easy to use, with instructions for use printed directly on the unit, making it ideal for use in education
  • Up to three samples can be viewed and tested at once
  • Supplied with spirit filled, mercury-free thermometer, which is not a health hazard if broken
  • BioCote® antimicrobial protection
Certifications: IP protection class according to DIN EN 60529: IP 20.
Ordering information: Supplied with spirit filled, mercury-free thermometer and a pack of 100 melting tubes (open both ends).