Melting point apparatus, digital, IA9000 series
Supplier: Electrothermal
621-1101EA 2520 EUR
621-1101 621-0022 621-1102
Melting point apparatus, digital, IA9000 series
Melting Point Apparatus
These Electrothermal microprocessor controlled melting point apparatuses allow quick and reliable testing of chemical samples. An adjustable extension arm facilitates comfortable viewing and helps reduce operator fatigue. The viewing head can be rotated for easier viewing and folds flat for easy and compact storage. Safety eyepiece reduces glare and protects your eyes from contact with the hot zone. Three audible beeps indicate when oven temperature has stabilised. Simple push-button controls are conveniently located. Heating chamber consists of aluminium heating block. RS232 output connector allows interface with the PR2000B printer or computer. Units can be calibrated in the field or can be sent back to the manufacturer for calibration.
Model IA9200:
Records up to four discrete temperatures by simply pushing the 'go-to' button
Date and time can be recorded or printed using the real time clock function
Model IA9300:
Used for recording beginning and ending points (melting point ranges) of each of three samples
Date and time can be recorded or printed using the real time clock function
LED indicator light identifies which of the three capillary tubes is being viewed or recorded