You Searched For: Magnifiers
44  results were found
SearchResultCount:"44"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: They provide high magnification for uniform illumination.
Supplier: VWR Collection
Description: Suitable for everyday applications.
Sale
Supplier: VWR Collection
Description: These illuminated desktop magnifiers, gooseneck or articulated arm type, include highly polished glass.
Sale
Catalog Number: (630-1947)
Supplier: VWR Collection
Description: These illuminated magnifiers can be fixed on a table or bench.
UOM: 1 * 1 items
Supplier: ESCHENBACH OPTIK GMBH
Description: These handheld folding magnifiers with classic design fit easily into any handbag or pocket.
Supplier: ESCHENBACH OPTIK GMBH
Description: Satin matt chrome plated brass frame with silicate glass lens.
Supplier: ESCHENBACH OPTIK GMBH
Description: LED illumination and coated lens on both sides with Ceratec® anti-scratch coating for optimised performances.
Supplier: ESCHENBACH OPTIK GMBH
Description: These pocket magnifiers provide magnification and illumination in a credit card format.
Catalog Number: (ESCH110910)
Supplier: ESCHENBACH OPTIK GMBH
Description: Aplanatic magnifier in a protective and robust case.
UOM: 1 * 1 items
Catalog Number: (630-1221)
Supplier: ESCHENBACH OPTIK GMBH
Description: Accessory for magnifier, varioLED, 2,5×, Ø: 132 mm
UOM: 1 * 1 items
Supplier: ESCHENBACH OPTIK GMBH
Description: These clip-on magnifiers are designed to provide hands-free magnification, and fit most eyeglass frames.
Supplier: ESCHENBACH OPTIK GMBH
Description: Swivelling black metallic magnifier with loop, lens frame made from black painted metal.
Supplier: ESCHENBACH OPTIK GMBH
Description: PXM® lightweight lenses with antistatic and scratch resistant coating. Ideal as a watchmaker's magnifier or gemstone magnifier.
Supplier: ESCHENBACH OPTIK GMBH
Description: Compact magnifiers with wide field of view and ultra-slim lenses.
Supplier: ESCHENBACH OPTIK GMBH
Description: These magnifiers featuring a modern design and a glass fibre reinforced PA body provide a distortion-free image.
Catalog Number: (630-1045)
Supplier: ESCHENBACH OPTIK GMBH
Description: Aspherical PXM® light-weight lens.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 44
no targeter for Bottom