You Searched For: Lights
134  results were found
SearchResultCount:"134"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Light Guide, Light guide focus, VWR®, For: Fibre optic light guides, Focus optic for fibre optic light guide
Supplier: VWR Collection
Description: Other optic versions (eg. flood or diffuse optics) or other colours or colour temperatures are available on request. Please contact your local Avantor...
Supplier: VWR Collection
Description: Halogen light, VWR®, Halogen bulb, 6 V/20 W
Supplier: VWR Collection
Description: For use with microscope series TL534B FL.
Supplier: VWR Collection
Description: The passively-cooled VisiLight® LED3 cold light source, with fanless cooling system, can be used for a variety of applications, such as quality assura...
Supplier: VWR Collection
Description: This xenon lamp is suitable for F-2500, F-2700, F-2710, F-4500, F-7000, F-2000, F-3000, F-3010, F-4000 and F-4010 spectrophotometers.
Supplier: VWR Collection
Description: For use with microscope series IT406/407.
Supplier: VWR Collection
Description: Spectrum lamp, Tungsten halogen lamp
Supplier: VWR Collection
Description: VisiLight® I-LED incident lights, with the unique 'modul-LED®' concept, makes it easy to adapt light colour and beam angle to any application simply b...
Catalog Number: (630-0858)
Supplier: VWR Collection
Description: Microscope fluorescent lamp 5 W for lower light, VWR®
UOM: 1 * 1 items
Supplier: VWR Collection
Description: The VisiLight® R-LED 66 ring light, with its innovative design, is adapted to precise lighting tasks. The 48 bright LED's provide homogeneous, flicker...
Supplier: Hach
Description: Spectrum lamp, Deuterium lamp
Supplier: SCHOTT AG Lighting and imaging
Description: Professional illumination for stereo microscopy. The VisiLED product line offers modular, homogeneous, and shadow-free LED illumination for stereo mic...
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: Ultraviolet light is considered an ancillary disinfectant to chemical disinfectants in the healthcare environment.
Catalog Number: (E90006)
Supplier: Biotium
Description: The PMA-Liteâ„¢ 2.0 LED photolysis device is a light-weight LED light box specifically designed for optimal photoactivation of samples treated with prop...
UOM: 1 * 1 items
Supplier: SCHOTT AG Lighting and imaging
Description: The EasyLED product line integrates advanced LED illumination with control functions in a single compact device. This saves space on the workbench and...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 134
no targeter for Bottom