You Searched For: UV Lights, Lamps and Bulbs
24  results were found
SearchResultCount:"24"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: Ultraviolet light is considered an ancillary disinfectant to chemical disinfectants in the healthcare environment.
Catalog Number: (SIMELD7220)
Supplier: Industrial Physics
Description: This robust, handheld rechargeable inspection spotlight is used to detect contaminations that react under UV-illumination and cannot be seen with nake...
UOM: 1 * 1 items
Catalog Number: (PIER95035)
Supplier: Thermo Fisher Scientific
Description: This compact ultraviolet lamp is intended for longwave, midrange and shortwave UV applications (252 nm, 302 nm and 365 nm). The lamp's innovative desi...
UOM: 1 * 1 items
Catalog Number: (135-6040)
Supplier: Thermo Scientific
Description: UV lamp, UV light option, factory installed
UOM: 1 * 1 items
Catalog Number: (SIMELD7290)
Supplier: Industrial Physics
Description: This UV pocket flashlight is designed to detect contaminations that react under UV illumination and can not be seen with naked eye such as some organi...
UOM: 1 * 1 items
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: UV lamp, Three wavelengths, UV lamp, 394×76×114 mm, 3UV™, 3UV-38, 8 W, 254/302/365 nm, W×D×H: 394×76×114 mm
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: Rechargeable 6-watt 26P series lamps are available in three dual-tube models. The lamps are equipped with an internal gel based, lead-acid battery, wh...
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: These lamps are excellent for quick field inspection or fluorescence applications.
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: UV lamp, UV bench lamp, shortwave, XX-20S, 20 W, 254 nm, W×D×H: 610×152×108 mm, Weight: 4,5 kg
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: UV field rack, With DC power supply, UV light field pack with DC power supply, two interchangeable light sources/wavelengths, light length: 44,5 mm, Pen-Ray®, 254/365 nm, Weight: 136 g
Supplier: CAMAG
Description: These UV lamps with plastic housing are designed for TLC applications.
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: Select from lamps with four, six or eight watts. Models available include 254 nm shortwave, 365 nm longwave, or a combination UV wavelengths, or UV an...
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: Made of durable, lightweight plastic, these compact 4 W UV lamps can be used as a handheld instrument or hands-free in combination with an optional st...
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: These lightweight lamps can be used with the optional lamp stand for hands-free operation. Replacement tubes are also available.
Supplier: CONSORT
Description: UV lamp, UV lamp with filter, UV lamp, 500×85×80 mm, 2×15 W, Intensity: 1800/1350 µW/cm², 312/365 nm, W×D×H: 500×85×80 mm
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: This lamp provides 100 W of high intensity, longwave (365 nm) illumination. It can be used for quality control, industrial inspection, and non-destruc...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 24
no targeter for Bottom