Mini incubator 4010
Supplier: GFL
390-8533EA 1390 EUR
GFLA4010
Mini incubator 4010
For thermostat control, heating and drying of test samples.
  • Microprocessor-control
  • Robust plexiglass lid for good visibility of contents
  • Quickly reaches the set temperature without overheating
Temperature range: Ambient + 8 to 60 ° C, Capacity 12 l