Thermostatic container, Oxitop® Box
Supplier: WTW
OxiTop®
705-1017EA 3377 EUR
705-1017
Thermostatic container, Oxitop® Box
Incubators Compact Benchtop Incubators
Benchtop container with a hinged transparent cover which can accommodate a maximum of either 12 OxiTop® simultaneous measurements or 20 Karlsruhe bottles. The compressor is CFC free and has an automatic defrosting system with condensate evaporation.
  • Temperature is fixed at 20 °C ±0,5 °C
  • Cross flow fan ensures uniform temperature distribution
  • Connection for an IS 6 or IS12 stirrer
H×W×D: 375×425×600 mm