You Searched For: Incubators
59  results were found
SearchResultCount:"59"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (390-0881)
Supplier: VWR Collection
Description: The IL 23R digital cooled incubator with Peltier technology offers an economical and space saving solution for microbiology applications at below room...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: IL 10, IL 23 and IL 56 digital incubators offer an economical and space saving solution for microbiology or haematology applications. The temperature ...
Supplier: VWR Collection
Description: Cooled incubators with Peltier technology feature excellent temperature stability and uniformity. They are energy efficient and environmentally friend...
Supplier: VWR Collection
Description: The ILCO 180 and ILCO 260 Premium offer optimal growth conditions for cell cultures and meet demanding requirements for best possible cell growth and ...
Sale
Supplier: VWR Collection
Description: INCU-Line® microbiological incubators are designed to cultivate organisms at homogeneous temperatures. They have a temperature range of 5 °C above amb...
Supplier: VWR Collection
Description: These compressor-cooled incubators provide a temperature range between –10 to +100 °C with forced air convection and fan speed control (10 to 100%). F...
Sale
Supplier: VWR Collection
Description: These compressor-cooled incubators provide a temperature range between 3 to 70 °C with forced air convection and fan speed control (50 to 100%). Featu...
Supplier: VWR Collection
Description: These compressor cooled incubators with forced air convection feature a stainless steel inner chamber and a powder coated sheet exterior. The IL 68R h...
Catalog Number: (390-0963)
Supplier: VWR Collection
Description: The INCU-Line® Tower is a general purpose, dual-chamber, cooled incubator with natural convection. This digitally cooled incubator with Peltier techno...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: These compressor-cooled incubators with forced air convection provide a temperature range between 3 to 70 °C. Featuring a stainless steel (DIN 1.4016)...
Supplier: Binder
Description: The KB series with compressor cooling consumes up to 30% less energy compared to its predecessor. Refrigerated incubators are mainly used for culturin...
Supplier: Binder
Description: The CBF series COâ‚‚ incubators are the premium series of COâ‚‚ incubators with humidity control. Units are suitable for all sensitive incubation applicat...
Supplier: Binder
Description: Avantgarde. Line incubators ensure stable and reliable incubation of microbiological cultures and guarantee reproducible results for routine tests in ...
Supplier: WTW
Description: These thermostatic cabinets are available in three sizes and different variants. Suited for BOD, OECD 301F, soil respiration and anaerobic degradatio...
Supplier: Binder
Description: COâ‚‚ incubators with a built in COâ‚‚ sensor which can be sterilised. Units offer optimum growth conditions for cell cultures and meet demanding requirem...
Supplier: Binder
Description: Thanks to thermoelectric cooling technology with heat dissipation, KB ECO cooled incubators are one of the most energy efficient cooling incubators on...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 59
no targeter for Bottom