Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (20)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
digital-thermo-hygrometers
https://ru.vwr.com/stibo/smallweb/std.lang.all/51/22/18945122.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"45"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   This wireless thermo-hygrometer, readable from up to 30 metres (433 MHz), can receive and display data from up to three remote modules. They can be co...
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   These thermo-hygrometers are ideal for routine measurements, monitoring 'round the clock', all quality control needs, and critical experimental requir...
Catalog Number: (620-1673)
Supplier:  VWR Collection
Description:   This thermometer/clock/humidity monitor shows time, temperature and humidity simultaneously. Internal sensors for use in hoods and storerooms.
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   Convenient triple-reading meter with 'always on' feature for continuous monitoring conditions. Audible and LED alarm may be set to trigger at three di...
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   This digital thermo-hygrometer continuously displays both relative humidity and temperature.
UOM:  1 * 1 items
Catalog Number: (620-1952)
Supplier:  VWR Collection
Description:   This accurate thermo-hygrometer with barometer features additional derived units: Dew point temperature, wet bulb temperature, moisture content and ab...
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   Digital thermo-hygrometer with probe, Traceable®, −40…+104 °C; 10 - 95 % RH , 89×140×32 mm
Catalog Number: (620-0915)
Supplier:  VWR Collection
Description:   This digital thermo-hygrometer is designed for temperature and humidity measurements in fridges and freezers. It is suitable for use in food and bever...
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 45
  1  2  3  4  5  6  Next