You Searched For: Homogenisers
126  results were found
SearchResultCount:"126"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Compact and easy to use blender, for sample volumes of 50 to 400 ml, efficiently blends solid or liquid samples in any sterile bags (plain or filter b...
Supplier: VWR Collection
Description: Pestles and microtubes manufactured from polybutylene terephthalate (PBTP) in a class 10000, ISO Class 7, M5.5 cleanroom. Designed for homogenising ce...
Supplier: VWR Collection
Description: Choice of aggregates for a wide range of laboratory applications from 5 ml up to 20 L. The VWR® Aggregates solve the standard problems that are faced ...
Sale
Supplier: VWR Collection
Description: Borosilicate glass tube, PTFE pestle with stainless steel shaft. Tissue grinders are used for controlled reduction of particle size and homogenisation...
Supplier: VWR Collection
Description: These compact homogenisers are suitable for all standard dispersing tasks and intensive mixing. The units are robust, and their stability prevents pul...
Sale
Catalog Number: (710-0953)
Supplier: VWR Collection
Description: Dispersing Element, Johnny sealing ring for blades, autoclavable, for use with Steri-Blostev 12 base fixing plate
UOM: 1 * 1 items
Catalog Number: (412-0167)
Supplier: VWR Collection
Description: A ball mill for disintregration of small sample volumes that can also be used to shake/mix microtubes and microplates. Wide range of applications in d...
UOM: 1 * 1 items
Catalog Number: (432-0366)
Supplier: VWR Collection
Description: The VWR® 4-place Mini Bead Mill homogeniser is compact and powerful.
UOM: 1 * 1 items
Catalog Number: (432-0367)
Supplier: VWR Collection
Description: The VWR® Bead Mill homogeniser is a part of the newest generation of Bead Mill homogenisers. This economical system is specifically designed for grind...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: VWR® Bead Mill MAX is the premier model in the VWR Bead Mill homogeniser range. VWR Bead Mill MAX is designed to grind, lyse or homogenise the most di...
Catalog Number: (710-0958)
Supplier: VWR Collection
Description: Sample volumes from 80 to 400 ml.
UOM: 1 * 1 items
Catalog Number: (710-2143)
Supplier: VWR Collection
Description: The VWR® Star Blender 400 Silent is a highly efficient laboratory blender for solid sample preparation. With a sterile bag, it guarantees optimal bacte...
UOM: 1 * 1 items
Catalog Number: (432-0274)
Supplier: Bertin Instruments
Description: Especially designed for low throughput laboratory workflows and personal use, Minilys® homogenises 3 samples simultaneously in 0,5/2 ml tubes or ...
UOM: 1 * 1 items
Catalog Number: (432-0478)
Supplier: Bertin Instruments
Description: Precellys® 24 Touch is the ideal laboratory homogeniser to prepare any kind of biological sample (animal and plant tissues, microorganisms) that can t...
UOM: 1 * 1 items
Supplier: KINEMATICA
Description: Dispersing aggregates available as standard (EC design) and special aggregates (W, B design).
Catalog Number: (431-0194)
Supplier: KINEMATICA
Description: Stand model, volume range 0,1 ml to 30 litres. The powerful PT 6100D was designed to deliver exactly reproducible homognising conditions for...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 126
no targeter for Bottom