Temperature regulator, MC242
Supplier: Electrothermal
454-0042EA 470 EUR
454-0042
Temperature regulator, MC242
Controllers Temperature Controllers
Single place percentage on/off temperature controller.
Recommended for use with CMU and cloth mantles.