Heating mantles with magnetic stirrer, Electromantle® EMA series
Supplier: Electrothermal
454-1417EA 1200 EUR
ELETEMA1000/CEB ELETEMA2000/CEB ELETEMA0100/CEB ELETEMA0250/CEB ELETEMA0500/CEB ELETEMA0050/CEB
Heating mantles with magnetic stirrer, Electromantle® EMA series
For round bottom flasks. Max. 450 °C, built-in electronic controller; stirring up to 520 min-1. Separate regulator and power-switch for heating and stirring.
  • Chemically resistant PP-housing
  • Bi-directional stirring
Delivery information: Supplied with a clamp for rod Ø 12,7 mm and one stirring bar.