Bunsen burner, FIREBOY eco
Supplier: Integra Biosciences
451-0136EA 625 EUR
451-0136
Bunsen burner, FIREBOY eco
Heaters Gas Burners
Mobile Bunsen burner for all applications where only foot switch ignition is sufficient and a timer is not needed. A connector for permanent gas supply is also included.
  • The flame remains ignited as long as the foot switch is activated
  • Two adjustment controls for gas and air, On/Off switch
W×D×H: 100×150×60 mm
Weight: 1,3 kg
Certifications: DVGW-approved
Delivery information: Includes 2 nozzles for propane/butane and natural gas, foot switch, power supply adapter and tubing adapter Ø 10 mm.