You Searched For: Gas Burners
73  results were found
SearchResultCount:"73"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (451-0156)
Supplier: USBECK Laborgeräte
Description: Nickel plated brass Bunsen burner ideal for heating or sterilising instruments and laboratory glassware.
UOM: 1 * 1 items
Supplier: USBECK Laborgeräte
Description: Meker burner with air regulation, suitable for all gases. With needle valve.
Supplier: USBECK Laborgeräte
Description: Nickel plated brass with cast iron foot.
Supplier: USBECK Laborgeräte
Description: Nickel plated brass with cast iron foot.
Supplier: USBECK Laborgeräte
Description: These air regulated bunsen burners have a double lever stop-cock with pilot flame inside.
Supplier: USBECK Laborgeräte
Description: Safety burners provide protection from the uncontrolled escape of gas in laboratories. The heat of the pilot flame generates an electric voltage withi...
Supplier: USBECK Laborgeräte
Description: This safety burner provides protection against uncontrolled escape of gas in laboratories. If the flame is extinguished, the gas supply is interrupted...
Supplier: USBECK Laborgeräte
Description: Nickel plated brass base, in powder coated grey.
Catalog Number: (USBE1240)
Supplier: USBECK Laborgeräte
Description: Burner for natural gas, adjustable. Maximum temperature of 1620 °C.
UOM: 1 * 1 items
Supplier: USBECK Laborgeräte
Description: DIN type.
Supplier: USBECK Laborgeräte
Description: Stainless steel burner for propane/butane mix. For temperatures up to 1400 °C.
Supplier: USBECK Laborgeräte
Description: Micro burner with air regulation and needle valve.
Catalog Number: (451-0089)
Supplier: USBECK Laborgeräte
Description: Nickel plated brass.
UOM: 1 * 1 items
Supplier: USBECK Laborgeräte
Description: Length: 155 mm.
Catalog Number: (451-0177)
Supplier: USBECK Laborgeräte
Description: Cartridge burner with air regulation. Maximum temperature of 1640 °C.
UOM: 1 * 1 items
Catalog Number: (451-0109)
Supplier: USBECK Laborgeräte
Description: Length: 220 mm.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 73
no targeter for Bottom