VWR gel dryers are available in two sizes and can accommodate gels up to 45×34 cm (MGD4534) and 50×40 cm (MGD5040). Each unit is light and robust, with the base unit manufactured in cast aluminium and the control panel in stainless steel. This ensures even heat distribution and resistance to corrosion in the base, while giving strength to the control panel. A clear cover allows the drying process to be observed. The units have two timers, one for temperature and one for the vacuum pump.
- Even heat distribution via built-in 800 Watt heating membrane
- Drying temperature can be set between ambient and 90 °C (±2 °C), and drying time set up to five hours in one minute intervals
- Vacuum pump (not included*) can be timed to automatically switch off after a set period of up to five hours
W×D×H: MGD4534 570×400×110; MGD5040 630×480×120 mm
*For details of a suitable pump please contact your local Avantor sales office.
Delivery information: Each gel dryer includes a stainless steel screen, a Mylar sheet, a porous PE sheet, and a clear silicone rubber overlay sheet.