Organic vapour monitors with back-up section
Supplier: 3M
151-0151EA 66.8 EUR
MMMA3520
Organic vapour monitors with back-up section
These organic vapour monitors use the proven scientific principle of diffusion, which involves the gradual mixing of molecules of two or more substances.
  • Monitoring compounds like acrylonitrile, butadiene and methylene chloride for which activated carbon shows limited capacity
  • Accuracy of ±25% at a 95% confidence level for workplace contaminants
  • 18 month shelf life
They are designed for measuring personal exposure in work areas of industries.