You Searched For: Freezers
41  results were found
SearchResultCount:"41"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: VWR® ULT Eco Premium freezers are designed for safe and reliable storage of sensitive samples. They feature ergonomic control and low energy consumpti...
Energy Efficient
Low Manufacturing Impact Sale
Catalog Number: (471-1312)
Supplier: VWR Collection
Description: Designed to meet the demands in daily laboratory use.
UOM: 1 * 1 items
New Product
Catalog Number: (471-1311)
Supplier: VWR Collection
Description: Designed to meet the demands in daily laboratory use, storage of biological products and storage of flammable materials.
UOM: 1 * 1 items
New Product
Supplier: Thermo Scientific
Description: Thermo Scientificâ„¢ TDE Series ultra-low temperature freezers, powered by H-drive, feature four upright models with 5 Inner doors, maximizing storage c...
Sustainable Materials
Catalog Number: (LIEBFNF4605)
Supplier: Liebherr
Description: Simple, classic, high-quality, with its shiny white-painted surface, the Liebherr fits particularly well into its surroundings and transcends fashion ...
UOM: 1 * 1 items
Supplier: REVCO TECHNOLOGIES
Description: Thermo Scientificâ„¢ TDE Series ultra-low temperature chest freezers set the standard for performance and reliable sample storage in heavy use situation...
Sustainable Materials
Supplier: GRANT INSTRUMENTS LTD.
Description: Controlled rate freezer and thawer that maximises cell survival without using liquid nitrogen.
New Product
Supplier: Thermo Scientific
Description: Thermo Scientific™ TDE Series ‒40 °C ultra-low temperature freezers, powered by H-drive, feature three upright models, maximizing storage capacity fro...
Sustainable Materials
Catalog Number: (LBCP7949030)
Supplier: LABCONCO
Description: FreeZone® benchtop shell freezers provide a compact system for prefreezing samples for lyophilization.
UOM: 1 * 1 items
Supplier: Liebherr
Description: Performance series with no lighting and perfection series with the LED front lighting provides optimal illumination of the drawers even in dark enviro...
Supplier: Liebherr
Description: These are heavy duty MediLine models with forced air cooling. The controller features door and temperature alarms which can be customised to suit the ...
Supplier: Liebherr
Description: Freezers designed for professional use. Reliably store non temperature-sensitive items even at ambient temperatures up to +40 °C with a constant cooli...
Supplier: Liebherr
Description: Freezers with a spark-free interior specifically for storing explosive and highly flammable substances in laboratories and in the chemical industry.
Supplier: Thermo Scientific
Description: The manual defrost, high-performance freezers are designed for applications in which even slight intermittent coil warming during auto defrost cannot ...
Energy Efficient
Safer Human and Environmental Health New Product
Supplier: Thermo Scientific
Description: Thermo Scientificâ„¢ TDE Series ultra-low temperature freezers, powered by H-drive, feature four upright models, maximizing storage capacity from 30000 ...
Sustainable Materials
Catalog Number: (471-1238)
Supplier: Stirling Ultracold
Description: Ultra-low temperature freezer featuring the free-piston Stirling engine. This next generation of environmentally sustainable ULT freezers achieve stab...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 41
no targeter for Bottom