Flocculators, SW5 and SW6
Supplier: Cole Parmer
709-0101EA 2000 EUR
STUASW5 STUASW6
Flocculators, SW5 and SW6
Designed for flocculation tests on water and effluent samples. The SW5 is a mini-version of the SW6 that accepts only two samples and is does not include the timer function. The SW6 is specifically designed for repeatable conditions between samples and from run to run. Two preset programmes allow routine speed and time parameters to be input and then selected by simply pressing one button. The intuitive touch sensitive keypad is totally flat for easy cleaning. Sample monitoring is assisted by base illumination against a black background.
  • Continuously variable stirring speed
  • Digital indication of time and speed
  • Two pre-set programmes
  • Built in illumination for easy viewing of samples
  • Timer feature controls stirring time and measures settling time
The SW5 is a mini-version of the SW6 that accepts only two samples and is does not include the timer function.
Ordering information: Beakers need to be ordered separately.