You Searched For: Soxhlet and Soxhlet-Type Extractors
34  results were found
SearchResultCount:"34"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Borosilicate 3.3 glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass. For extraction from solids with solvents.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, Borosilicate glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, extractor head only.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Soxhlet Extractor, LF 100 flange, Extraction cap.: 2000 ml, Flask capacity: - ml
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass.
Catalog Number: (KAVA632442108090)
Supplier: BOHEMIA CRISTAL HANDELSGES.MBH
Description: Borosilicate glass.
UOM: 1 * 1 items
Catalog Number: (KAVA632442101100)
Supplier: BOHEMIA CRISTAL HANDELSGES.MBH
Description: Borosilicate glass. For extraction from solids with solvents.
UOM: 1 * 1 items
Supplier: GLASWARENFABRIK KARL HECHT
Description: Borosilicate glass.
Supplier: BUCHI
Description: Borosilicate glass.
Supplier: SciLabware
Description: PYREX®, borosilicate glass.
Supplier: BEHR
Description: Complete compact extraction system with basic rack, heating device, holder, hoses and glassware (reaction flasks, extractor, Dimroth condenser for ext...
Catalog Number: (BEHRB00218435)
Supplier: BEHR
Description: Behr equipment for Soxhlet extraction comply with the most various requirements of daily laboratory routines.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 34
no targeter for Bottom