Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Parallel Synthesis Systems
https://ru.vwr.com/stibo/smallweb/std.lang.all/96/32/18989632.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"5"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (ASYNADS3-PS-17.4)
Supplier:  ASYNT
Description:   The DrySyn parallel synthesis kit allows the scientist to heat and stir up to 27 small scale reactions in parallel using any standard hotplate.
UOM:  1 * 1 KIT
Supplier:  BÜCHI
Description:   The Syncore® Analyst offers fast, gentle and environmentally friendly multiple sample concentration to a predefined volume.
Supplier:  BÜCHI
Description:   This compact instrument for parallel evaporation of up to six samples under vacuum conditions is designed to optimise existing personal process workfl...
Supplier:  BÜCHI
Description:   Highly modular parallel evaporator with interchangeable racks for 4 to 96 samples, covering working volumes of 0,5 to 500 ml. Due to a number of optio...
Supplier:  CEM
Description:   Make higher purity peptides faster. Microwave energy drives biochemical reactions to completion up to 10 times faster than by conventional methods, re...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 5  of 5