You Searched For: Evaporators
39  results were found
SearchResultCount:"39"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: This is a basic chemical laboratory instrument suitable for evaporation, distillation and separation of chemicals. It can be used as a multifunctional...
Sale
Supplier: VWR Collection
Description: This is an essential instrument for use in chemical laboratories for efficient and gentle removal of solvents from samples by evaporation. It offers a...
Supplier: LABCONCO
Description: These systems use vacuum along with heat and vortex motion to help speed evaporation and provide an alternative to traditional centrifugal evaporation...
Supplier: Heidolph Instruments GmbH & Co.KG
Description: Hei-VAP Expert Control rotary evaporators allow central control of all process parameters: Vacuum, cooling temperature, rotation and heating bath temp...
Supplier: LABCONCO
Description: This system uses nitrogen blow down, vortex motion and heat to speed evaporation of up to 48 small samples or eight 450 milliliter samples at once.
Sale
Supplier: LABCONCO
Description: CentriVap® vacuum concentrators are available in a range of configurations. The CentriVap® DNA model is supplied with a built-in 34 litres/min diaphra...
Supplier: Heidolph Instruments GmbH & Co.KG
Description: These evaporators are equipped with proven smart details such as Easy-Clip, patented clamping sleeve, lift stop to limit immersion depth and universal...
Supplier: LABCONCO
Description: CentriVap® vacuum concentrators are available as complete systems or system packages. The acid resistant model is recommended for corrosive solvents (...
Supplier: LABCONCO
Description: Combine the separate concentrator and cold trap components into one space-saving console and include casters for portability from laboratory to labora...
Supplier: LABCONCO
Description: The refrigerated CentriVap centrifugal concentrator is designed to rapidly concentrate multiple small heat-sensitive samples, such as RNA and proteins...
Supplier: LABCONCO
Description: This evaporator speeds evaporation of up to 50 samples at the same time using dry heat and nitrogen blow down. Unlike water bath heaters, the dry bloc...
Supplier: LABCONCO
Description: An efficient, automatic evaporator system which can be used with a broad range of aggressive chemicals. In RapidVap Nâ‚‚ systems, a stream of nitrogen o...
Supplier: Heidolph Instruments GmbH & Co.KG
Description: Large scale evaporators up to 20 L.
Catalog Number: (535-0107)
Supplier: LABCONCO
Description: The CentriVap micro IR vacuum concentrator has a very small footprint and therefore fits in tight spaces. It is ideal for small samples such as DNA an...
UOM: 1 * 1 items
Supplier: THERMO SAVANT EC
Description: The Thermo Scientificâ„¢ Savantâ„¢ SpeedVacâ„¢ SPD210 vacuum concentrator is a large-capacity vacuum concentrator with a modular design, ideal for drying aq...
Supplier: IKA
Description: The RV 3 rotary evaporator is the ideal entry-level model from the IKA rotary evaporator portfolio. It is suitable for a multitude of uses in the chem...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 39
no targeter for Bottom