Electro cell manipulator, ECM® 2001
Supplier: BTX
732-0617EA 0 EUR
BTXI45-0018 BTXI45-0080 BTXI45-0010 BTXI45-0011 BTXI45-0012
Electro cell manipulator, ECM® 2001
The ECM® 2001 is a multifunctional electro cell manipulation and electrofusion instrument. It generates an AC waveform for benign dielectrophoretic alignment of cells. With microsecond switch-over time from AC to DC, efficient fusion can take place. After fusion AC re-application keeps cells together for the rounding off process. The DC square pulse section can be used as a stand-alone electroporation system with a wide range of voltages and pulse lengths capable of operating into low impedence loads.
  • Versatile system suitable for electro cell fusion, embryo manipulation and/or electroporation
  • Microprocessor controlled precision
  • Wide range of parameters
* Includes ECM® 2001 generator, disposable cuvettes (1 mm, 2 mm and 4 mm gap sizes), safety stand and cuvette rack.
** Includes ECM® 2001 generator, microslides (10 each with 0,5 mm, 1,0 mm and 1 each with 3,2 mm gap), micrograbber cables and coaxial banana plug cables.
*** Includes ECM® 2001 generator, microslides (10 each with 0,5 mm and 1 each with 3,2 mm gap), micrograbber cables, meander fusion chamber (0,2 mm gap on glass microslide, including connector), flat electrode/divergent field (1,0 mm gap, 0,5 ml divergent field geometry), electrode adapter, coaxial banana plug cable.
****Includes ECM 2001 generator, microslides, rectangular wire (3.2 mm gap, 10 mm gap), adapter set, micrograbber adapter cables, cytofusion medium C, 500 ml, safety stand for flatpack.