Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (5)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Electrophoresis Power Supplies
https://ru.vwr.com/stibo/smallweb/std.lang.all/51/83/18945183.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"17"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   VWR® electrophoresis power supplies are ideal for most general electrophoresis applications including DNA, RNA and protein separations and Western blo...
Supplier:  VWR Collection
Description:   VWR® Power Source power supplies meet a wide range of applications, including DNA, RNA and protein electrophoresis, and blotting. Designed with safety...
Supplier:  CONSORT
Description:   A complete range of power supplies for electrophoresis applications, including DNA sequencing, flat bed iso-electric focussing, horizontal and vertica...
Supplier:  CONSORT
Description:   Recommended for mini horizontal and vertical gels.
Supplier:  CONSORT
Description:   A complete range of power supplies for electrophoresis applications, including DNA sequencing, flat bed iso-electric focussing, horizontal and vertica...
Catalog Number: (732-2381)
Supplier:  HOEFER ELECTROPHORESIS
Description:   The midrange PS600 power supply is designed for electrophoresis and blotting techniques, including large format and high throughput applications. The ...
UOM:  1 * 1 items
Catalog Number: (732-1693)
Supplier:  HOEFER ELECTROPHORESIS
Description:   The PS200HC 200 V high current power supply is ideal for large electrophoretic blotting applications and for low voltage electrophoresis runs. The sma...
UOM:  1 * 1 items
Catalog Number: (CBSCEPS-300X)
Supplier:  Expedeon
Description:   N/A
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 17
  1  2  3  Next