You Searched For: Electrochemical Meters
196  results were found
SearchResultCount:"196"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: These precise instruments measure dissolved oxygen in % and ppm (mg/l) and temperature. A built-in microprocessor stores, calculates, and compensates ...
Catalog Number: (662-2283)
Supplier: VWR Collection
Description: This handheld measuring device features an ergonomic design, dust and waterproof housing in accordance with IP 65/67 and an illuminated display. This ...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: These compact, portable meters measure dissolved oxygen and temperature using a smart optical sensor. Values are transmitted from and to the sensor in...
Sale
Supplier: VWR Collection
Description: These economical, robust and waterproof pH meters are supplied in complete, ready to use kits that incorporate a robust carry case, electrode and temp...
Sale
Supplier: VWR Collection
Description: These digital meters combine six parameters in one instrument: pH, mV, temperature, conductivity, dissolved oxygen and turbidity. IDP technology optim...
Catalog Number: (663-0359)
Supplier: VWR Collection
Description: These portable, economical meters can simultaneously measure conductivity, specific conductance, salinity, TDS (Total Dissolved Solids) and temperatur...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Simple, compact instruments designed for quick accurate results in an economical pen format. Ideal for agriculture, education, fruit and vegetable was...
Sale
Catalog Number: (662-2380)
Supplier: VWR Collection
Description: The user friendly instrument is distinguished by a large a 127 mm colour graphic touch screen. The meter can be used for accurate measurements of pH, ...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: This user friendly portable instrument with robust IP 67 rated housing provides high resolution and accuracy for precise measurements of pH, mV, condu...
Catalog Number: (663-0234)
Supplier: VWR Collection
Description: This user friendly portable instrument with robust IP 67 rated housing provides high resolution and accuracy for precise measurements in field applica...
UOM: 1 * 1 SET
Catalog Number: (664-0177)
Supplier: VWR Collection
Description: This user friendly portable instrument with robust IP 67 rated housing provides high resolution and accuracy for precise optical oxygen measurements i...
UOM: 1 * 1 SET
Catalog Number: (664-0175)
Supplier: VWR Collection
Description: This user friendly instrument with articulated electrode stand and IP 43-rated housing provides high resolution and accuracy for precise measurem...
UOM: 1 * 1 SET
Catalog Number: (664-0236)
Supplier: VWR Collection
Description: These precise instruments measure dissolved oxygen in % and ppm (mg/L) and temperature. A built-in microprocessor stores, calculates and compensates f...
UOM: 1 * 1 items
Catalog Number: (663-0349)
Supplier: VWR Collection
Description: This high resolution device for measuring the conductivity up to 100 μS/cm is supplied with a stainless steel measuring cell for measurements in pure ...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: User friendly instrument distinguished by its large 127 mm colour graphic touch screen. The meter can be used for accurate measurements of conductivit...
Supplier: VWR Collection
Description: User friendly instrument, with articulated electrode stand and IP 43-rated housing, provides high resolution and accuracy for precise multi-param...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 196
no targeter for Bottom