Bottle-top dispensers, Dispensette® III
Supplier: Brand
Dispensette® III
613-3306EA 0 EUR
613-3306 BRND4701161 BRND4701350 BRND4701341 613-3308 613-3310
Bottle-top dispensers, Dispensette® III
Bottle Top Dispensers
Fixed volume, analogue-adjustable or Digital Easy Calibration.
  • SafetyPrime™ valve (optional) for recirculation of reagent during priming
  • Three different models of volume adjustment available: Fixed (Fixed-volume for standard applications; simple calibration adjustment with supplied tool) and Analog (Analog volume display, fast volume adjustment; simple calibration adjustment with supplied tool)
  • Digital EASY CALIBRATION: Easy to read digital display; dispensing volume can be set accurately and reproducibly
  • Easy Calibration technique for adjustment in seconds without tools; alteration of factory setting is automatically indicated
  • DE-M marking
  • Easy to calibrate and adjust in order to comply with ISO 9001 and GLP guidelines; alteration of factory setting is automatically indicated
For dispensing aggressive reagents such as concentrated acids (e.g. H₃PO₄, H₂SO₄), alkalis (e.g. NaOH, KOH), saline solutions, and many organic solvents. Parts that are in contact with the liquid are made from borosilicate glass, ceramic, platinum-iridium, ETFE, FEP, PFA, PTFE and PP (discharge tube safety screw cap). Colour code: Red.
* Special fixed volumes: 0,5 to 100 ml (please state when ordering)!
Ordering information: Supplied with discharge tube, telescopic filling tube, mounting tool, PP adapters, and certificate of performance. SafetyPrime™ recirculation valve (optional).