You Searched For: Data Loggers
78  results were found
SearchResultCount:"78"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (620-2694)
Supplier: VWR Collection
Description: TraceableLIVE® provides simple, digital monitoring of CO₂, temperature and humidity in greenhouses, micro labs, biofuel research and other environment...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: TraceableGO Bluetooth temperature and humidity dataloggers fill in the unknowns between destinations. Bluetooth device is the ideal transit partner fo...
Supplier: VWR Collection
Description: These data logging thermometers with Ethernet offer an easy and secure way to connect your smartphone, tablet or personal computer remotely with your ...
Supplier: VWR Collection
Description: Easily access collected data between destinations with Bluetooth® data loggers.
Catalog Number: (620-1965)
Supplier: VWR Collection
Description: This stand-alone data logger records readings on a removable SD card. The card reader (supplied) plugs into any USB computer port for an instant repor...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: These loggers travel with shipments to track relative humidity and temperature. For complete flexibility, the user-defined recording interval may be s...
Supplier: VWR Collection
Description: This temperature logger can display, record and report data during storage, handling and transportation of products.
Catalog Number: (SENA0096647494944)
Supplier: SenseAnywhere
Description: The SenseAnywhere AccessPoint is an economic plug and play device that connects SenseAnywhere’s wireless Sensors to the Cloud service of SenseAnywhere...
UOM: 1 * 1 items
Supplier: SenseAnywhere
Description: Accurate, maintenance-free wireless temperature data logger for stationary or transport use. Perfect for high humidity environments with rapid tempera...
Supplier: EBRO
Description: The EBI 11 mini data loggers are considered as the perfect solution for temperature and pressure measurements in tight spaces.
Supplier: EBRO
Description: These temperature and humidity loggers record data automatically. The LCD shows the measured value, battery level and unit of measure. These waterproo...
Sale
Supplier: EBRO
Description: EBI 25 data logging system consists of a data logger, an interface and evaluation and monitoring software. It can be used in the food industry and lab...
Supplier: SenseAnywhere
Description: The SenseAnywhere AiroSensor X ER model 20-20-43 is like the most popular AiroSensor T ER model 20-20-31 but with an IP 67 class connector featuring t...
Catalog Number: (SENA0096647495002)
Supplier: SenseAnywhere
Description: The SenseAnywhere 4 - 20 mA input module is a smart piece of technology that enables digital measurements for all analogue products with a 4 - 20 mA o...
UOM: 1 * 1 items
Supplier: EBRO
Description: EBI 25-TX for wireless monitoring of temperature measurements ensures that perishable goods are produced and stored at the right conditions at all tim...
Catalog Number: (TEST0563.4406)
Supplier: Testo
Description: Airflow Combokit 1 equipped with clearly structured measurement menu for volume flow and parallel determination of air velocity, differential pressure...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 78
no targeter for Bottom