You Searched For: Colorimeters
30  results were found
SearchResultCount:"30"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: These portable colorimeters are designed for precise water analysis.
Supplier: Hach
Description: The DR300 colorimeter is a reliable instrument that provides state-of-the-art data transfer capability and connection to Claros. Simple, intuitive ope...
Supplier: Hach
Description: Generic pocket colorimeter DR300 instruments are intended for customised applications not included in standard analysis systems. They are available in...
Supplier: Hach
Description: The LICO 620 and LICO 690 colorimeters are ideal for professional colour measurements of clear, transparent liquids with reference beam technology.
Supplier: WATER-ID PRIMELAB
Description: The next generation of multitest Photometer, with pH - EC - TDS - ORP - Temperature measurement functionality.
New Product
Catalog Number: (705-0744)
Supplier: Hach
Description: The DR900 is a robust multiparameter configured for immediate use.
UOM: 1 * 1 items
Catalog Number: (ALEXADI-907-030A)
Supplier: Enzo Life Sciences
Description: The creatinine colorimetric detection kit utilises a single step liquid detection reagent that is safer and less time consuming than other assay metho...
UOM: 1 * 1 KIT
Supplier: Lovibond Tintometer
Description: Accessory for colorimeter, Automatic, transmittance, Optical glass cell 33 mm path length
Supplier: Lovibond Tintometer
Description: The RT300, RT400 and RT500 are versatile, portable spectrophotometers in a choice of diffuse/8° sphere or 0/45° optical geometry that are designed to ...
Supplier: Lovibond Tintometer
Description: Available for many of the commonly used colour scales, these kits are a convenient means of ordering the complete range of equipment required. Each sy...
Supplier: Lovibond Tintometer
Description: A series of spectrophotometric colorimeters for colour grading of optically clear samples. Each version of the PFXi 195 includes a selection of...
Supplier: Lovibond Tintometer
Description: The PFX995 spectrophotometers are top of the range, versatile instruments that offer comprehensive colour data requirements for either t...
Supplier: Lovibond Tintometer
Description: The Lovibond® EComparator series provides an easy way to transition from subjective visual measurement to a non subjective, accurate electronic measur...
Supplier: Lovibond Tintometer
Description: The Lovibond® System 3000 are a series of single-scale, 3-field instruments for visually determining the colour of samples by direct comparison with c...
Supplier: Lovibond Tintometer
Description: A series of automatic spectrophotometric colorimeters for objective colour analysis of edible oils, chemicals, petrochemicals, fuels and other transpa...
Supplier: Lovibond
Description: Lovibond® Comparator system 2000+ is an extremely versatile, modular system for testing water. The battery operated portable lighting unit can be int...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 30
no targeter for Bottom