You Searched For: Light Boxes
6  results were found
SearchResultCount:"6"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (E90006)
Supplier: Biotium
Description: The PMA-Liteâ„¢ 2.0 LED photolysis device is a light-weight LED light box specifically designed for optimal photoactivation of samples treated with prop...
UOM: 1 * 1 items
Catalog Number: (733-2003)
Supplier: EDVOTEK EUROPE LTD
Description: The Edvotek White Light Box features a spacious 25×25 cm viewing area illuminated by long life LEDs and is housed in a slim aluminium body. It is desi...
UOM: 1 * 1 items
Catalog Number: (HEAT120661)
Supplier: HEATHROW
Description: This small light box saves bench space and can be stored in a drawer.
UOM: 1 * 1 items
Supplier: KAISER FOTOTECHNIK GMBH & CO.K
Description: Super flat and dimmable daylight light box with build-in rechargeable battery.
Catalog Number: (ADLM6031511)
Supplier: ADELPHI MANUFACTURING
Description: Designed for use in pharmaceutical and laboratory environments, the Apollo II Liquid Viewer for transparent glass containers provides a simple method ...
UOM: 1 * 1 items
Catalog Number: (ADLM6031520)
Supplier: ADELPHI MANUFACTURING
Description: Designed for use in pharmaceutical and laboratory environments, the Apollo II Liquid Viewer for coloured or opaque glass containers provides a simple ...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 6 of 6
no targeter for Bottom