You Searched For: Colony Counters
21  results were found
SearchResultCount:"21"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (710-2246)
Supplier: VWR Collection
Description: The MCC 2000 is designed for the rapid and accurate counting of bacterial colonies.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Premium 150 and Premium 90 HR colony counters and zone measurements systems.
Sale
Supplier: VWR Collection
Description: Counter-Pen is a combination marker and digital counter designed for the laboratory and plant. Each time a black mark is made with the felt-tipped pen...
Catalog Number: (710-2072)
Supplier: VWR Collection
Description: Auto lite is the latest affordable, automated colony counting device. The Auto lite is available in one configuration and features simpl...
UOM: 1 * 1 items
Catalog Number: (710-1868)
Supplier: INTERSCIENCE
Description: The Scan 100 is designed with a LED system and an ergonomic hand rest.
UOM: 1 * 1 items
Catalog Number: (INSC437000)
Supplier: INTERSCIENCE
Description: High resolution colony counter that can be linked to a PC via a FireWire connection.
UOM: 1 * 1 items
Catalog Number: (INSC438000)
Supplier: INTERSCIENCE
Description: Scan 4000 is an ultra-high definition automatic colony counter and inhibition zone reader for high resolution colour reading of colonies and inhibitio...
UOM: 1 * 1 items
Catalog Number: (INSC436300)
Supplier: INTERSCIENCE
Description: Automatic colony counter with a digital camera and high technology software.
UOM: 1 * 1 items
Catalog Number: (INSC436000)
Supplier: INTERSCIENCE
Description: High technology colony counter with software that can be linked to a PC via a FireWire connection.
UOM: 1 * 1 items
Catalog Number: (REIC13332600)
Supplier: REICHERT INC.
Description: This manual colony counter is used in counting bacteria colonies that are presented against a dark background positioned to eliminate parallax errors.
UOM: 1 * 1 items
Catalog Number: (720-0098)
Supplier: GERBER FUNKE DR N.
Description: Customising the colour and the brightness makes work easy. A total of 65 colour variations are available using the primary colours. The pressure sensi...
UOM: 1 * 1 items
Catalog Number: (710-0732)
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: The innovative and compact design of the ColonyDoc-Itâ„¢ enables automated, fast and accurate colony counting. The high resolution DSLR colour camera is...
UOM: 1 * 1 items
Catalog Number: (710-1970)
Supplier: SYNBIOSIS
Description: aCOLyte 3 HD uses a powerful algorithm for colony counting on pour plates and Spiral® plates.
UOM: 1 * 1 items
Catalog Number: (710-1700)
Supplier: Bel-Art Products, a Part of SP
Description: The display with backlight for easy viewing in low light conditions. The audible alert can be turned off if desired, and the counting function can be ...
UOM: 1 * 1 items
Supplier: Cole Parmer
Description: Versatile unit can be used with 50 to 90 mm Petri dishes.
Catalog Number: (SYNBACOLAD2)
Supplier: SYNBIOSIS
Description: aCOLade2 is designed with an antimicrobial protection. The pressure of marking a colony with a felt-tip pen registers a cumulative count. The pressure...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 21
no targeter for Bottom