You Searched For: Chart Recorders
12  results were found
SearchResultCount:"12"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (HERA6383-6)
Supplier: Thermo Scientific
Description: These recorders are ideal when ultra-low temperature freezers need to meet validation and regulatory requirements.
UOM: 1 * 1 items
Catalog Number: (SIMEAB3500)
Supplier: Industrial Physics
Description: Drying time recorder tool can be used as an optional test tool for the automatic film applicators. This device allows the user to accurately define th...
UOM: 1 * 1 items
Catalog Number: (REVC6185)
Supplier: REVCO TECHNOLOGIES
Description: Chart for recorders, Chart paper, for -86 and -40 °C freezers, +50…−115 °C, Hours (24) or Days (7)
UOM: 1 * 1 items
Supplier: JULES , RICHARD INSTRUMENTS
Description: Chart for recorder, For graphic recorders, +35…-15 °C
Catalog Number: (JULS00297)
Supplier: JULES , RICHARD INSTRUMENTS
Description: Chart for recorders, Diagram sheets for visiotambour range of measurement –15 +40 °C, 7 days
UOM: 1 * 100 items
Catalog Number: (620-0432)
Supplier: DICKSON
Description: These are single use logger accurately records temperatures from start to finish of transport and documents adherence to critical storage environments...
UOM: 1 * 1 items
Supplier: DICKSON
Description: Chart for recorder, C314, +10...+35 °C
Supplier: DICKSON
Description: These 6" charts are designed for 6" chart recorders.
Supplier: DICKSON
Description: These self-contained temperature recorders on a Ø 100 mm disc provide a permanent record of temperatures with an average response time of 11 minutes t...
Supplier: DICKSON
Description: These recorders are used to record temperature simply and reliably. They provide the users with immediate access to data and suit applications in manu...
Catalog Number: (620-1484)
Supplier: DICKSON
Description: The vaccine chart recorder can be placed right inside the refrigerator or freezer along with the vaccines.
UOM: 1 * 60 items
Supplier: LAMBRECHT
Description: Chart for recorder, –10…+50 °C
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 12 of 12
no targeter for Bottom