Microcentrifuges, high speed, ventilated/refrigerated, CT15E / CT15RE
Supplier: VWR Hitachi
521-3601EA 3890 EUR
HITC905607V1 HITC905606V1
Microcentrifuges, high speed, ventilated/refrigerated, CT15E / CT15RE
High technology benchtop centrifuges, ventilated or refrigerated, designed for standard applications, as well as biosafety or molecular biology applications.
  • Exceptionally low noise
  • Compact and safe - door interlock, dual overspeed detector, imbalance detector, abnormal motor temperature detector, automatic door closure system
  • Easy to use front panel
  • Autoclavable rotors (T15A62 and T15A63 rotors can also be used for biosafety applications)
Certifications: Compliance with CE marking requirements for mechanical and electrical safety, and electromagnetic compatibility (61010-1, 61010-2-020, EN 61326)
Ordering information: Supplied with T15A61 fixed angle rotor, 24×2,0 ml, 21 500 ×g, and plastic lid. T15A62 and T15A63 rotors available separately.