You Searched For: Benchtop Centrifuges
47  results were found
SearchResultCount:"47"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: These versatile centrifuges are ideal for a range of routine applications in pharmaceutical and biotechnology industries, as well as for academic rese...
Supplier: VWR Collection
Description: These versatile centrifuges are ideal for a range of routine applications in pharmaceutical and biotechnology industries, as well as for academic rese...
Supplier: VWR Collection
Description: These small bench top centrifuges are ideal for research and clinical applications in pharmaceutical and biotechnical industries, as well as for acade...
Sale
Catalog Number: (521-3345)
Supplier: VWR Collection
Description: The CompactStar CS8 is a small ventilated bench top centrifuge designed for the separation of various samples in the fields of chemistry, life science...
UOM: 1 * 1 items
Catalog Number: (521-2853)
Supplier: VWR Collection
Description: The CompactStar CS4 centrifuge is designed for the separation of various samples in the fields of chemistry, life science, clinical research and routi...
UOM: 1 * 1 items
Supplier: Thermo Scientific
Description: Thermo Scientific™ Megafuge ST1/ST1R Plus bench top centrifuges offer intuitive controls and versatility for power at every turn.
Sale
Supplier: Thermo Scientific
Description: These durable centrifuges are available as ventilated or refrigerated versions and provide exceptional capacity in a compact design with a smart, simp...
Supplier: Thermo Scientific
Description: Process a range of general-purpose applications including cell culture, microplate, bioproduction, blood separation, and microbiology with the Thermo ...
Supplier: Thermo Scientific
Description: Ideal for routine processing, these centrifuges combine exceptional capacity, ergonomics, and value with market-leading capabilities including unparal...
Supplier: THARMAC GmbH
Description: Versatile cytocentrifuges suitable for small and large sample volumes, designed for thin layer and mono layer cell preparations in medical application...
Supplier: THARMAC GmbH
Description: The cytology specialist. Large selection of open and closed rotors - also compatible with Shandon™ Cytospin™ systems. Optional rapid quick-change roto...
Catalog Number: (512-0714)
Supplier: THARMAC GmbH
Description: The genius among the centrifuges. Automatic rotor detection and speed adjustment on max. 2000 rpm when using cytorotors.
UOM: 1 * 1 items
Supplier: Thermo Scientific
Description: Ergonomically designed, the Thermo Scientific™ Multifuge X1/X1R Pro series of bench top centrifuges have a touch screen interface that enables easier ...
Sale
Supplier: Thermo Scientific
Description: Process a range of general purpose applications including cell culture, microplate, bioproduction, blood separation and microbiology with the Thermo S...
Catalog Number: (521-4098)
Supplier: Beckman Coulter
Description: With its low sound levels, you will hardly notice this new assistant despite it racing through run cycles at up to 150000 min⁻¹ (2500 revolutions per ...
UOM: 1 * 1 items
Catalog Number: (521-3365)
Supplier: Thermo Scientific
Description: General Purpose Pro Centrifuges Cell Therapy Systems (CTS™) Series consist of equipment, documentation, and certifications, as well as compliance serv...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 47
no targeter for Bottom