VWR®, Semi-dry blotter, 20×20 cm
Supplier: VWR
700-7156EA 588.61 EUR
700-7156
VWR®, Semi-dry blotter, 20×20 cm
Blotting Apparatus
This unit is manufactured from rugged acrylic components and built to comply with all statutory safety regulations. One electrode is manufactured from stainless steel and the other from platinised titanium, therefore making them virtually corrosion-free. This prevents deposits being placed on the membrane and the electrodes will last almost indefinitely, as they will not deteriorate. The unit is designed to produce a uniform system for the transfer of proteins and nucleic acids.
  • Very low buffer volume and current (0,8 mA per cm²) of the gel produces very little heat, preventing band distortion
  • Transfers can be achieved in less than an hour depending on the molecular weight of the molecules to transfer
  • Either single gels or multiple gel stacks can be used
Unit W×L×H: 275×265×65 mm