VWR®, Stainless Steel Sand Baths
Supplier: VWR
VWR®
460-3230EA 372 EUR
460-3230 460-3231 460-3232 460-3233 460-3227 460-3228 460-3229
VWR®, Stainless Steel Sand Baths
Heaters Sandbaths
Ideal for irregular-shaped vessels. Stainless steel construction for superior corrosion resistance. Designed to hold sand, stainless steel shot or non volatile liquids.